Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
bcar1
Entrez Gene ID
447979
Gene Name
BCAR1, Cas family scaffold protein
Gene Aliases
CASB3b, Crkas, cas, p130cas
Chromosome Location
Chr.4: 51199060 - 51289113 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126064.1 NP_001119536.1 4 - 5 1201 110
XM_012960799.2 XP_012816253.1 4 - 5 3008 110
GenBank mRNA BC076945.1 - 4 - 5 1465 110
BC166334.1 - 4 - 5 1201 110

View Full Product Details