Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ube2e2
Entrez Gene ID
100144938
Gene Name
ubiquitin conjugating enzyme E2E 2
Gene Aliases
ubch8, ube2e2-a, ube2e2-b
Chromosome Location
Chr.6: 77679445 - 77807354 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126511.1 NP_001119983.1 3 - 4 553 82
GenBank mRNA BC158231.1 - 3 - 4 553 82

View Full Product Details