Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem86a
Entrez Gene ID
100145092
Gene Name
transmembrane protein 86A
Gene Aliases
-
Chromosome Location
Chr.7: 105905403 - 105910083 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126611.1 NP_001120083.1 1 - 2 81 72
XM_018095257.1 XP_017950746.1 2 - 3 419 72
GenBank mRNA BC158523.1 - 1 - 2 81 72

View Full Product Details