Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hunk
Entrez Gene ID
100145703
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
mak, makv
Chromosome Location
Chr.2: 11393910 - 11430238 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127077.1 NP_001120549.1 2 - 3 1025 92
GenBank mRNA BC161479.1 - 2 - 3 1025 92

View Full Product Details