Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cfap53
Entrez Gene ID
100145337
Gene Name
cilia and flagella associated protein 53
Gene Aliases
ccdc11
Chromosome Location
Chr.1: 191937023 - 191948430 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126809.1 NP_001120281.1 5 - 6 1078 68
GenBank mRNA BC160546.1 - 5 - 6 1078 68
CU025145.1 - 6 - 7 1129 68

View Full Product Details