Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpatch3
Entrez Gene ID
100145580
Gene Name
G-patch domain containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 51979653 - 51994229 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126999.1 NP_001120471.1 5 - 6 1214 81
GenBank mRNA BC161307.1 - 5 - 6 1214 81

View Full Product Details