Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mettl17
Entrez Gene ID
100145785
Gene Name
methyltransferase like 17
Gene Aliases
mett11d1
Chromosome Location
Chr.1: 130982400 - 130998793 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127148.1 NP_001120620.1 10 - 11 998 96
GenBank mRNA BC161764.1 - 10 - 11 998 96
CU075325.1 - 10 - 11 1002 96

View Full Product Details