Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100145817
Entrez Gene ID
100145817
Gene Name
uncharacterized LOC100145817
Gene Aliases
il-2
Chromosome Location
Chr.1: 56639938 - 56644338 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018091006.1 XP_017946495.1 4 - 5 365 93
GenBank mRNA EF513165.1 - 2 - 3 178 93

View Full Product Details