Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hmgxb3
Entrez Gene ID
100147705
Gene Name
HMG-box containing 3
Gene Aliases
hbcx, hmgx3, smf
Chromosome Location
Chr.3: 6155672 - 6204495 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127168.1 NP_001120640.1 12 - 13 2420 86
XM_012958613.2 XP_012814067.1 13 - 14 2585 86
XM_012958614.2 XP_012814068.1 13 - 14 2511 86
GenBank mRNA BC166189.1 - 12 - 13 2420 86

View Full Product Details