Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fzd3
Entrez Gene ID
100038082
Gene Name
frizzled class receptor 3
Gene Aliases
Fz-3, Xfz3, frizzled-3, frizzled3, frz-3, frz3, fz3, hfz3
Chromosome Location
Chr.Un|NW_016683402: 418623 - 467307 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129910.1 NP_001123382.1 - 1809 103
GenBank mRNA BC124561.1 - - 1766 103
BC167603.1 - - 1809 103

View Full Product Details