Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
myl10
Entrez Gene ID
100170437
Gene Name
myosin light chain 10
Gene Aliases
mylc2pl
Chromosome Location
Chr.2: 42241791 - 42281788 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130210.1 NP_001123682.1 3 - 4 188 64
XM_012953918.1 XP_012809372.1 3 - 4 240 64
GenBank mRNA BC167140.1 - 3 - 4 188 64

View Full Product Details