Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpat4
Entrez Gene ID
100170573
Gene Name
glycerol-3-phosphate acyltransferase 4
Gene Aliases
agpat6, agpat6.2, lpaat-zeta, lpaatz, tsarg7
Chromosome Location
Chr.3: 119911450 - 119924627 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130350.1 NP_001123822.1 8 - 9 1543 101
XM_012958656.2 XP_012814110.1 9 - 10 1701 101
XM_012958657.2 XP_012814111.1 9 - 10 1728 101
GenBank mRNA BC167630.1 - 8 - 9 1543 101

View Full Product Details