Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
irf2bpl
Entrez Gene ID
100170574
Gene Name
interferon regulatory factor 2 binding protein-like
Gene Aliases
c14orf4, eap1, irf2bp1, irf2bpl-a, irf2bpl-b
Chromosome Location
Chr.8: 61441699 - 61445231 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130351.1 NP_001123823.1 1 - 1 2148 98
GenBank mRNA BC167631.1 - 1 - 1 2148 98

View Full Product Details