Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trappc4
Entrez Gene ID
779542
Gene Name
trafficking protein particle complex 4
Gene Aliases
cgi-104, hspc172, ptd009, sbdn, trs23
Chromosome Location
Chr.7: 85207029 - 85216983 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142905.2 NP_001136377.1 2 - 3 362 74
XM_018095427.1 XP_017950916.1 2 - 3 748 74
GenBank mRNA BC121584.1 - 2 - 3 297 74
BC152700.1 - 2 - 3 337 74
BC166938.1 - 2 - 3 363 74
CU075488.1 - 2 - 3 359 74

View Full Product Details