Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sp7
Entrez Gene ID
100189548
Gene Name
Sp7 transcription factor
Gene Aliases
osx
Chromosome Location
Chr.2: 144214865 - 144231732 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135118.1 NP_001128590.1 2 - 2 754 110
GenBank mRNA AB236908.1 - 2 - 2 754 110

View Full Product Details