Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sut4
Entrez Gene ID
100240688
Gene Name
sucrose transporter4
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144014.1 NP_001137486.1 - 1366 73
GenBank mRNA AY603492.1 - - 1366 73
AY639018.1 - - 1446 73

View Full Product Details