Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100283810
Entrez Gene ID
100283810
Gene Name
ATPP2-B12
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001156708.1 NP_001150180.1 - 444 73
GenBank mRNA EU966026.1 - - 444 73

View Full Product Details