Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
glk1
Entrez Gene ID
541882
Gene Name
G2-like1
Gene Aliases
ZEAMMB73_644019
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111548.1 NP_001105018.1 - 1245 57
GenBank mRNA AF318580.1 - - 1245 57

View Full Product Details