Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pdc3
Entrez Gene ID
541919
Gene Name
pyruvate decarboxylase3
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111582.1 NP_001105052.1 - 1450 77
GenBank mRNA AF370005.1 - - 1450 77
AY106187.1 - - 1504 77
BT056069.1 - - 1490 77
D14457.1 - - 140 77
EU970907.1 - - 1423 77
Z21722.1 - - 140 77

View Full Product Details