Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
zds1
Entrez Gene ID
542604
Gene Name
zeta carotene desaturase candidate
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112139.1 NP_001105609.1 - 1233 99
GenBank mRNA AF047490.1 - - 1233 99
AY110101.1 - - 1233 99

View Full Product Details