Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
gst1
Entrez Gene ID
542366
Gene Name
glutathione-S-transferase1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111942.1 NP_001105412.1 - 273 62
GenBank mRNA AY103561.1 - - 281 63
M16901.1 - - 210 62
X06754.1 - - 273 62

View Full Product Details