Gene Transcripts
- Gene Symbol
- Speer4cos
- Entrez Gene ID
- 75858
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 4c, opposite strand transcript
- Gene Aliases
- 4930568L21Rik, SPEER-7, Speer7-ps, Speer7-ps1
- Chromosome Location
- Chr. 5: 15680710 - 15714596 on Build GRCm38
- UniGene ID
- Mm.359463
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001585.3 |
|
1 |
88 |
|
View Full Product Details