Gene Transcripts

Gene Symbol
Ceacam-ps1
Entrez Gene ID
100038912
Gene Name
carcinoembryonic antigen-related cell adhesion molecule pseudogene 1
Gene Aliases
Gm1978
Chromosome Location
Chr. 7: 16649030 - 16657478 on Build GRCm38
UniGene ID
Mm.389813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003247.2 3 564
GenBank mRNA CK023612.1 - 562


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CK022275.1 - 3 564 Chr. 7 - 16648934 - 16657478 [-]
GenBankmRNA CK022560.1 - 3 553 Chr. 7 - 16648940 - 16657467 [-]
GenBankmRNA CK023298.1 - 3 561 Chr. 7 - 16648954 - 16657476 [-]

View Full Product Details