Gene Transcripts
- Gene Symbol
- Ppifos
- Entrez Gene ID
- 68173
- Gene Name
- peptidylprolyl isomerase F, opposite strand
- Gene Aliases
- 4930520F12Rik, Ppifs
- Chromosome Location
- Chr. 14: 25696356 - 25701282 on Build GRCm38
- UniGene ID
- Mm.444727
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028021.1 |
|
3 |
1118 |
|
GenBank mRNA |
AK019686.1 |
- |
3 |
1120 |
|
View Full Product Details