Gene Transcripts
- Gene Symbol
- Nphs1os
- Entrez Gene ID
- 445267
- Gene Name
- nephrosis 1 homolog, nephrin, opposite strand
- Gene Aliases
- Nphs1as
- Chromosome Location
- Chr. 7: 30462164 - 30465591 on Build GRCm38
- UniGene ID
- Mm.398140
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004443.1 |
|
3 |
954 |
|
GenBank mRNA |
AY422730.1 |
- |
3 |
954 |
|
View Full Product Details