Gene Transcripts
- Gene Symbol
- Mir7-1
- Entrez Gene ID
- 723902
- Gene Name
- microRNA 7-1
- Gene Aliases
- Mirn7-1, mir-7a-1, mmu-mir-7-1
- Chromosome Location
- Chr. 13: 58392779 - 58392886 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029825.1 |
|
1 |
23 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA881786.1 |
- |
1 |
74 |
|
Chr. 13 - 58392382 - 58392937 [-] |
View Full Product Details