Gene Transcripts
- Gene Symbol
- Mir24-2
- Entrez Gene ID
- 723960
- Gene Name
- microRNA 24-2
- Gene Aliases
- Mirn24-2, mir-24-2
- Chromosome Location
- Chr. 8: 84208815 - 84208921 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029741.1 |
|
1 |
60 |
|
View Full Product Details