Gene Transcripts

Gene Symbol
Dctd
Entrez Gene ID
320685
Gene Name
dCMP deaminase
Gene Aliases
6030466N05Rik
Chromosome Location
Chr. 8: 48099092 - 48141667 on Build GRCm38
UniGene ID
Mm.121549
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027759.1 5 916
GenBank mRNA AK138614.1 - 5 704
AK171894.1 - 5 713
BB193146.2 - 3 485
BQ572929.1 - 3 390

View Full Product Details