Gene Transcripts
- Gene Symbol
- Psg-ps1
- Entrez Gene ID
- 232919
- Gene Name
- pregnancy specific glycoprotein pseudogene 1
- Gene Aliases
- AV036018, Cea-6, Cea6
- Chromosome Location
- Chr. 7: 17672313 - 17682060 on Build GRCm38
- UniGene ID
- Mm.153893
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002857.1 |
|
5 |
1562 |
|
GenBank mRNA |
AK132281.1 |
- |
5 |
1562 |
|
M83348.1 |
- |
5 |
1558 |
|
View Full Product Details