Gene Transcripts
- Gene Symbol
- 5830417I10Rik
- Entrez Gene ID
- 100302730
- Gene Name
- gon-4-like pseudogene
- Gene Aliases
- 3632445O20Rik, AA387005
- Chromosome Location
- Chr. 3: 88777057 - 88829371 on Build GRCm38
- UniGene ID
- Mm.482358
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028359.1 |
|
18 |
7132 |
|
GenBank mRNA |
AK031583.1 |
- |
1 |
150 |
|
AK049055.1 |
- |
3 |
1573 |
|
View Full Product Details