Gene Transcripts

Gene Symbol
AFG3L1P
Entrez Gene ID
172
Gene Name
AFG3 like matrix AAA peptidase subunit 1, pseudogene
Gene Aliases
AFG3, AFG3L1
Chromosome Location
Chr. 16: 89972580 - 90000787 on Build GRCh38
UniGene ID
Hs.534773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003226.1 6 715
NR_003227.1 6 642
NR_003228.1 6 681
GenBank mRNA AF329691.1 - 5 466
AF329693.1 - 5 466
AF329694.1 - 4 397
AK056488.1 - 6 668
AK091293.1 - 6 678
AK097044.1 - 6 629
AK126911.1 - 5 606
AK128804.1 - 3 254
BC021245.2 - 4 288


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB001266.1 - 4 439 Chr. 16 - 89972586 - 89984591 [+]
GenBankmRNA BF974129.1 - 289 Chr. - - [-]
GenBankmRNA BG570268.1 - 6 589 Chr. 16 - 89972597 - 89984754 [+]
GenBankmRNA BX114649.1 - 3 664 Chr. 16 - 89979841 - 89984629 [+]
GenBankmRNA BQ893592.1 - 3 564 Chr. 16 - 89979940 - 89990996 [+]
GenBankmRNA DA316906.1 - 4 442 Chr. 16 - 89972586 - 89990886 [+]

View Full Product Details