Gene Transcripts
- Gene Symbol
- OR7E91P
- Entrez Gene ID
- 79315
- Gene Name
- olfactory receptor family 7 subfamily E member 91 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 2: 71024075 - 71029931 on Build GRCh38
- UniGene ID
- Hs.327033
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002185.3 |
|
1 |
107 |
|
GenBank mRNA |
BC014374.1 |
- |
1 |
110 |
|
BC030991.1 |
- |
1 |
89 |
|
View Full Product Details