Gene Transcripts
- Gene Symbol
- FAM66A
- Entrez Gene ID
- 100133172
- Gene Name
- family with sequence similarity 66 member A
- Gene Aliases
- Chromosome Location
- Chr. 8: 12362019 - 12411001 on Build GRCh38
- UniGene ID
- Hs.721640
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026789.1 |
|
4 |
699 |
|
GenBank mRNA |
AK311040.1 |
- |
4 |
700 |
|
View Full Product Details