Gene Transcripts
- Gene Symbol
- LINC01551
- Entrez Gene ID
- 387978
- Gene Name
- long intergenic non-protein coding RNA 1551
- Gene Aliases
- C14orf23, c14_5148
- Chromosome Location
- Chr. 14: 28772704 - 28794794 on Build GRCh38
- UniGene ID
- Hs.633254
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB464055.1 |
- |
1 |
24 |
|
Chr. 14 - 28772795 - 28813287 [+] |
GenBankmRNA |
BX419086.2 |
- |
1 |
23 |
|
Chr. 14 - 28772796 - 28792615 [+] |
View Full Product Details