Gene Transcripts
- Gene Symbol
- SCARNA4
- Entrez Gene ID
- 677771
- Gene Name
- small Cajal body-specific RNA 4
- Gene Aliases
- ACA26
- Chromosome Location
- Chr. 1: 155925958 - 155926086 on Build GRCh38
- UniGene ID
- Hs.678903
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003005.1 |
|
1 |
15 |
|
GenBank mRNA |
AJ609483.1 |
- |
1 |
15 |
|
View Full Product Details