Gene Transcripts

Gene Symbol
A1BG-AS1
Entrez Gene ID
503538
Gene Name
A1BG antisense RNA 1
Gene Aliases
A1BG-AS, A1BGAS, NCRNA00181
Chromosome Location
Chr. 19: 58351970 - 58355183 on Build GRCh38
UniGene ID
Hs.709582
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015380.2 4 1950
GenBank mRNA AK027222.1 - 4 1950
BC006144.1 - 1 782
BC040926.1 - 3 1538

View Full Product Details