Gene Transcripts

Gene Symbol
FOXN3-AS2
Entrez Gene ID
29018
Gene Name
FOXN3 antisense RNA 2
Gene Aliases
PRO1768
Chromosome Location
Chr. 14: 89576216 - 89577477 on Build GRCh38
UniGene ID
Hs.673124
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024620.1 1 1213
GenBank mRNA AF118074.1 - 1 1213
BC069658.1 - 1 1158
BC069757.1 - 1 1162
BC069775.1 - 1 1162

View Full Product Details