Gene Transcripts
- Gene Symbol
- RNU6ATAC
- Entrez Gene ID
- 100151684
- Gene Name
- RNA, U6atac small nuclear (U12-dependent splicing)
- Gene Aliases
- RNU6ATAC1, U6ATAC
- Chromosome Location
- Chr. 9: 134164440 - 134164564 on Build GRCh38
- UniGene ID
- Hs.545589
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023344.1 |
|
1 |
104 |
|
GenBank mRNA |
U62823.1 |
- |
1 |
104 |
|
View Full Product Details