Gene Transcripts
- Gene Symbol
- ASAP1-IT1
- Entrez Gene ID
- 29065
- Gene Name
- ASAP1 intronic transcript 1
- Gene Aliases
- ASAP1-IT, ASAP1IT, ASAP1IT1, DDEF1IT1, HSPC054, NCRNA00050
- Chromosome Location
- Chr. 8: 130295355 - 130296533 on Build GRCh38
- UniGene ID
- Hs.639318
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002765.2 |
|
1 |
427 |
|
View Full Product Details