Gene Transcripts
- Gene Symbol
- ALDH1L2
- Entrez Gene ID
- 160428
- Gene Name
- aldehyde dehydrogenase 1 family member L2
- Gene Aliases
- mtFDH
- Chromosome Location
- Chr. 12: 105019784 - 105084609 on Build GRCh38
- UniGene ID
- Hs.42572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027752.1 |
|
4, 5 |
724 |
|
GenBank mRNA |
BC103935.1 |
- |
4, 5 |
619 |
|
View Full Product Details