Gene Transcripts
- Gene Symbol
- CXADRP3
- Entrez Gene ID
- 440224
- Gene Name
- coxsackie virus and adenovirus receptor pseudogene 3
- Gene Aliases
- Chromosome Location
- Chr. 18: 14477952 - 14498706 on Build GRCh38
- UniGene ID
- Hs.721675
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024076.1 |
|
1 |
95 |
|
- Gene Symbol
- LINC01297
- Entrez Gene ID
- 106146148
- Gene Name
- long intergenic non-protein coding RNA 1297
- Gene Aliases
- LA16c-83F12.6
- Chromosome Location
- Chr. 22: 15746674 - 15778289 on Build GRCh38
- UniGene ID
- Hs.435289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY338954.1 |
- |
3 |
4991 |
|
View Full Product Details