Gene Transcripts
- Gene Symbol
- SNORA58
- Entrez Gene ID
- 677836
- Gene Name
- small nucleolar RNA, H/ACA box 58
- Gene Aliases
- ACA58, SNORA58B
- Chromosome Location
- Chr. 3: 131478998 - 131479233 on Build GRCh38
- UniGene ID
- Hs.735762
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002985.2 |
|
1 |
28 |
|
GenBank mRNA |
AJ609450.1 |
- |
1 |
28 |
|
DW436402.1 |
- |
1 |
44 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DW421138.1 |
- |
1 |
44 |
|
Chr. 3 - 131478953 - 131479236 [-] |
View Full Product Details