Gene Transcripts
- Gene Symbol
- HAR1A
- Entrez Gene ID
- 768096
- Gene Name
- highly accelerated region 1A (non-protein coding)
- Gene Aliases
- HAR1F, LINC00064, NCRNA00064
- Chromosome Location
- Chr. 20: 63101292 - 63104385 on Build GRCh38
- UniGene ID
- Hs.549606
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003244.1 |
|
|
1 |
925 |
|
GenBank mRNA |
DQ860409.1 |
- |
LIT3337 HAR1F |
1 |
925 |
|
View Full Product Details