Gene Transcripts
- Gene Symbol
- DEFA10P
- Entrez Gene ID
- 449493
- Gene Name
- defensin alpha 10, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 8: 6968141 - 6969113 on Build GRCh38
- UniGene ID
- Hs.552564
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029386.1 |
|
1 |
131 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AY746435.1 |
- |
1 |
131 |
|
Chr. 8 - 6968141 - 6969113 [-] |
View Full Product Details