Gene Transcripts
- Gene Symbol
- SNORA13
- Entrez Gene ID
- 654322
- Gene Name
- small nucleolar RNA, H/ACA box 13
- Gene Aliases
- ACA13
- Chromosome Location
- Chr. 5: 112161485 - 112161617 on Build GRCh38
- UniGene ID
- Hs.689709
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002922.1 |
|
1 |
62 |
|
GenBank mRNA |
AJ609455.1 |
- |
1 |
62 |
|
View Full Product Details