Gene Transcripts
- Gene Symbol
- GBP1P1
- Entrez Gene ID
- 400759
- Gene Name
- guanylate binding protein 1 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 89407679 - 89424934 on Build GRCh38
- UniGene ID
- Hs.661968
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003133.2 |
|
5 |
1061 |
|
GenBank mRNA |
AK309962.1 |
- |
5 |
1061 |
|
View Full Product Details