Gene Transcripts
- Gene Symbol
- BREA2
- Entrez Gene ID
- 286076
- Gene Name
- breast cancer estrogen-induced apoptosis 2
- Gene Aliases
- Chromosome Location
- Chr. 8: 143697115 - 143698413 on Build GRCh38
- UniGene ID
- Hs.178095
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015445.1 |
|
1 |
133 |
|
GenBank mRNA |
AL833948.1 |
- |
1 |
133 |
|
AY940073.1 |
- |
1 |
1102 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000531565.1 |
- |
1 |
1094 |
|
Chr. 8 - 143696154 - 143698413 [+] |
View Full Product Details