Gene Transcripts

Gene Symbol
FKBP9P1
Entrez Gene ID
360132
Gene Name
FK506 binding protein 9 pseudogene 1
Gene Aliases
FKBP9L
Chromosome Location
Chr. 7: 55681074 - 55704567 on Build GRCh38
UniGene ID
Hs.446691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003949.1 6 1220
NR_027339.1 4 1324
NR_027340.1 3 953
NR_027342.1 4 1234
GenBank mRNA BC011872.2 - 4 1234

View Full Product Details