Gene Transcripts
- Gene Symbol
- OR6W1P
- Entrez Gene ID
- 89883
- Gene Name
- olfactory receptor family 6 subfamily W member 1 pseudogene
- Gene Aliases
- OR6W1, sdolf
- Chromosome Location
- Chr. 7: 143062284 - 143063785 on Build GRCh38
- UniGene ID
- Hs.339818
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002140.1 |
|
1 |
203 |
|
View Full Product Details