Gene Transcripts
- Gene Symbol
- EMBP1
- Entrez Gene ID
- 647121
- Gene Name
- embigin pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 121519112 - 121571888 on Build GRCh38
- UniGene ID
- Hs.697682
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003955.1 |
|
8 |
3169 |
|
GenBank mRNA |
AK128714.1 |
- |
8 |
3170 |
|
View Full Product Details